COMH93D5E440 Combi-Blocks, Inc.

4-(Chloromethyl)-1-(4-fluorophenyl)-1h-1,2,3-triazole

  • MDL number MFCD16748031
  • Cas Number : 1250623-38-1
  • Molecular Weight : 211.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6052

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6052
Availability details for
Quantity
  • Availability
  • Add to Cart