COMH93D5E469 Combi-Blocks, Inc.

1-([3-Chloro-4-(trifluoromethoxy)phenyl]methyl)piperazine

  • MDL number MFCD16811222
  • Cas Number : 1240565-36-9
  • Molecular Weight : 294.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6096

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6096
Availability details for
Quantity
  • Availability
  • Add to Cart