COMH93D5E46E Combi-Blocks, Inc.

(2-Aminoethyl)(tert-butyl)amine dihydrochloride

  • MDL number MFCD16817515
  • Cas Number : 149374-23-2
  • Molecular Weight : 189.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6101

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6101
Availability details for
Quantity
  • Availability
  • Add to Cart