COMH93D5E473 Combi-Blocks, Inc.

(R)-1,1,1-Trifluoro-2-propylamine

  • MDL number MFCD16817687
  • Cas Number : 779303-24-1
  • Molecular Weight : 113.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6106

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6106
Availability details for
Quantity
  • Availability
  • Add to Cart