COMH93D5E477 Combi-Blocks, Inc.

1-(3-Bromophenyl)pentylamine

  • MDL number MFCD16822180
  • Cas Number : 1249620-29-8
  • Molecular Weight : 242.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6110

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6110
Availability details for
Quantity
  • Availability
  • Add to Cart