COMH93D5E48C Combi-Blocks, Inc.

N-(Pyrimidin-5-ylmethyl)cyclohexanamine

  • MDL number MFCD16864834
  • Cas Number : 1339341-94-4
  • Molecular Weight : 191.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6135

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6135
Availability details for
Quantity
  • Availability
  • Add to Cart