COMH93D5E4AA Combi-Blocks, Inc.

5-Fluoro-3-methyl-2,3-dihydro-1h-inden-1-one

  • MDL number MFCD16876796
  • Cas Number : 61241-54-1
  • Molecular Weight : 164.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6171

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6171
Availability details for
Quantity
  • Availability
  • Add to Cart