COMH93D5E51F Combi-Blocks, Inc.

(7-Chloro-2,3-dihydrofuro[2,3-c]pyridin-3-yl)methanol

  • MDL number MFCD17012758
  • Cas Number : 174469-04-6
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6312

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6312
Availability details for
Quantity
  • Availability
  • Add to Cart