COMH93D5E53E Combi-Blocks, Inc.

4-Oxopiperidine-2-carboxylic acid hydrochloride

  • MDL number MFCD17015054
  • Cas Number : 99979-55-2
  • Molecular Weight : 179.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6352

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6352
Availability details for
Quantity
  • Availability
  • Add to Cart