COMH93D5E598 Combi-Blocks, Inc.

Methyl 4-(cyanomethyl)thiophene-2-carboxylate

  • MDL number MFCD17215389
  • Cas Number : 878741-67-4
  • Molecular Weight : 181.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6470

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6470
Availability details for
Quantity
  • Availability
  • Add to Cart