COMH93D5E60F Combi-Blocks, Inc.

Ethyl 5-hydroxy-3,6-dihydro-2h-pyran-4-carboxylate

  • MDL number MFCD17676281
  • Cas Number : 324767-92-2
  • Molecular Weight : 172.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6612

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6612
Availability details for
Quantity
  • Availability
  • Add to Cart