COMH93D5E6A8 Combi-Blocks, Inc.

(S)-N-(2,3-Dihydro-1h-inden-1-yl)-7h-pyrrolo[2,3-d]pyrimidin-4-amine

  • MDL number MFCD18207034
  • Cas Number : 905580-86-1
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6802

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6802
Availability details for
Quantity
  • Availability
  • Add to Cart