COMH93D5E6AA Combi-Blocks, Inc.

(R)-1-((Benzyloxy)carbonyl)-2-methylpyrrolidine-2-carboxylic acid

  • MDL number MFCD18207152
  • Cas Number : 63399-74-6
  • Molecular Weight : 263.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6804

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6804
Availability details for
Quantity
  • Availability
  • Add to Cart