COMH93D5E6B6 Combi-Blocks, Inc.

tert-Butyl 4-(3-nitropyridin-4-yl)piperazine-1-carboxylate

  • MDL number MFCD18207662
  • Cas Number : 608142-93-4
  • Molecular Weight : 308.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6821

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6821
Availability details for
Quantity
  • Availability
  • Add to Cart