COMH93D5E6BD Combi-Blocks, Inc.

7-Fluoro-1-oxo-2,3-dihydro-1h-indene-5-carbonitrile

  • MDL number MFCD18208657
  • Cas Number : 1260017-98-8
  • Molecular Weight : 175.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6829

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6829
Availability details for
Quantity
  • Availability
  • Add to Cart