COMH93D5E6F7 Combi-Blocks, Inc.

Lac beta(1-4)lac-beta-c10

  • MDL number MFCD18252875
  • Cas Number : 1858224-00-6
  • Molecular Weight : 806.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6900

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6900
Availability details for
Quantity
  • Availability
  • Add to Cart