COMH93D5E6F9 Combi-Blocks, Inc.

3-Bromo-9-(2-ethylhexyl)-9h-carbazole

  • MDL number MFCD18252906
  • Cas Number : 628336-85-6
  • Molecular Weight : 358.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6902

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6902
Availability details for
Quantity
  • Availability
  • Add to Cart