COMH93D5E701 Combi-Blocks, Inc.

3-Bromo-7-methoxy-1,5-naphthyridine

  • MDL number MFCD18253954
  • Cas Number : 1246550-41-3
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6913

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-6913
Availability details for
Quantity
  • Availability
  • Add to Cart