COMH93D5E7D6 Combi-Blocks, Inc.

4-Bromo-2-methyl-1h-indole-6-carboxylic acid

  • MDL number MFCD18381252
  • Cas Number : 1260382-87-3
  • Molecular Weight : 254.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7183

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7183
Availability details for
Quantity
  • Availability
  • Add to Cart