COMH93D5E7FA Combi-Blocks, Inc.

2-Bromo-6-formylbenzonitrile

  • MDL number MFCD18390711
  • Cas Number : 77532-87-7
  • Molecular Weight : 210.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7226

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7226
Availability details for
Quantity
  • Availability
  • Add to Cart