COMH93D5E807 Combi-Blocks, Inc.

2-(3-Bromo-5-methoxyphenyl)acetonitrile

  • MDL number MFCD18391933
  • Cas Number : 123018-27-9
  • Molecular Weight : 226.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7246

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7246
Availability details for
Quantity
  • Availability
  • Add to Cart