COMH93D5E820 Combi-Blocks, Inc.

5-(Difluoromethyl)-2-fluorobenzonitrile

  • MDL number MFCD18395398
  • Cas Number : 934175-58-3
  • Molecular Weight : 171.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7276

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7276
Availability details for
Quantity
  • Availability
  • Add to Cart