COMH93D5E82E Combi-Blocks, Inc.

3-(3-Fluoro-4-trifluoromethoxy-phenyl)-propionic acid

  • MDL number MFCD18397097
  • Cas Number : 1261616-38-9
  • Molecular Weight : 252.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7295

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7295
Availability details for
Quantity
  • Availability
  • Add to Cart