COMH93D5E848 Combi-Blocks, Inc.

Methyl 2-methoxy-3-methylphenylacetate

  • MDL number MFCD18399542
  • Cas Number : 1261827-91-1
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7326
Availability details for
Quantity
  • Availability
  • Add to Cart