COMH93D5E866 Combi-Blocks, Inc.

3-Hydroxy-5-(trifluoromethyl)quinoline

  • MDL number MFCD18414815
  • Cas Number : 1261678-80-1
  • Molecular Weight : 213.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7362
Availability details for
Quantity
  • Availability
  • Add to Cart