COMH93D5E88B Combi-Blocks, Inc.

Fmoc-ser(tbu)-thr(psime,mepro)-oh

  • MDL number MFCD18427361
  • Cas Number : 1266350-99-5
  • Molecular Weight : 524.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7407

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7407
Availability details for
Quantity
  • Availability
  • Add to Cart