COMH93D5E89C Combi-Blocks, Inc.

Methyl 2-formyl-6-nitrobenzoate

  • MDL number MFCD18432396
  • Cas Number : 142314-70-3
  • Molecular Weight : 209.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7428

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7428
Availability details for
Quantity
  • Availability
  • Add to Cart