COMH93D5E8E3 Combi-Blocks, Inc.

9H-Carbazole-2-carboxylic acid, ethyl ester

  • MDL number MFCD18452378
  • Cas Number : 82408-83-1
  • Molecular Weight : 239.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7515

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7515
Availability details for
Quantity
  • Availability
  • Add to Cart