COMH93D5E94D Combi-Blocks, Inc.

(S)-3-Cyclopropylmorpholine

  • MDL number MFCD18665146
  • Cas Number : 1270264-56-6
  • Molecular Weight : 127.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7652

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7652
Availability details for
Quantity
  • Availability
  • Add to Cart