COMH93D5E9D5 Combi-Blocks, Inc.

2-Pyridinecarboxaldehyde,6-ethenyl-

  • MDL number MFCD18816459
  • Cas Number : 579500-16-6
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7851

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7851
Availability details for
Quantity
  • Availability
  • Add to Cart