COMH93D5E9EF Combi-Blocks, Inc.

2-Fluoro-6-isopropylaniline

  • MDL number MFCD18824824
  • Cas Number : 126476-48-0
  • Molecular Weight : 153.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7886

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7886
Availability details for
Quantity
  • Availability
  • Add to Cart