COMH93D5EA4C Combi-Blocks, Inc.

Ethyl 2-amino-2-(pyridin-3-yl)acetate hydrochloride

  • MDL number MFCD18909148
  • Cas Number : 1439903-29-3
  • Molecular Weight : 216.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7999

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-7999
Availability details for
Quantity
  • Availability
  • Add to Cart