COMH93D5EA8D Combi-Blocks, Inc.

5-Methoxyquinolin-6-amine

  • MDL number MFCD18971257
  • Cas Number : 54620-48-3
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8088

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8088
Availability details for
Quantity
  • Availability
  • Add to Cart