COMH93D5EAAC Combi-Blocks, Inc.

4-(2-Methoxyethoxy)-3-(trifluoromethyl)benzoic acid

  • MDL number MFCD19132769
  • Cas Number : 1271093-46-9
  • Molecular Weight : 264.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8123

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8123
Availability details for
Quantity
  • Availability
  • Add to Cart