COMH93D5EAC3 Combi-Blocks, Inc.

(1R,2R,3S)-2-Aminocyclohexane-1,3-diol

  • MDL number MFCD19203876
  • Cas Number : 2753-42-6
  • Molecular Weight : 131.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8154

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8154
Availability details for
Quantity
  • Availability
  • Add to Cart