COMH93D5EAD1 Combi-Blocks, Inc.

3-Methylbutane-1,3-diamine

  • MDL number MFCD19207215
  • Cas Number : 116473-67-7
  • Molecular Weight : 102.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8174

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8174
Availability details for
Quantity
  • Availability
  • Add to Cart