COMH93D5EAD7 Combi-Blocks, Inc.

3-Amino-2-fluoropropan-1-ol

  • MDL number MFCD19211115
  • Cas Number : 78542-46-8
  • Molecular Weight : 93.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8181

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8181
Availability details for
Quantity
  • Availability
  • Add to Cart