COMH93D5EAEF Combi-Blocks, Inc.

N-Methyl-1,2,4-thiadiazol-5-amine

  • MDL number MFCD19218125
  • Cas Number : 100703-97-7
  • Molecular Weight : 115.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8209

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8209
Availability details for
Quantity
  • Availability
  • Add to Cart