COMH93D5EB0A Combi-Blocks, Inc.

1,2-Dimethylcyclopropanecarboxylic acid

  • MDL number MFCD19228404
  • Cas Number : 594846-52-3
  • Molecular Weight : 114.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8245

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8245
Availability details for
Quantity
  • Availability
  • Add to Cart