COMH93D5EB0E Combi-Blocks, Inc.

4-Methylcyclohexa-1,4-diene-1-carboxylic acid

  • MDL number MFCD19228627
  • Cas Number : 53312-53-1
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8251

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8251
Availability details for
Quantity
  • Availability
  • Add to Cart