COMH93D5EB3E Combi-Blocks, Inc.

4-Fluorobenzyl piperidine-4-carboxylate

  • MDL number MFCD19323041
  • Cas Number : 1267245-27-1
  • Molecular Weight : 237.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8315

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8315
Availability details for
Quantity
  • Availability
  • Add to Cart