COMH93D5EB4C Combi-Blocks, Inc.

3,7-Dimethylbenzo[d]oxazol-2(3h)-one

  • MDL number MFCD19374514
  • Cas Number : 1267216-41-0
  • Molecular Weight : 163.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8330

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8330
Availability details for
Quantity
  • Availability
  • Add to Cart