COMH93D5EB56 Combi-Blocks, Inc.

Du-14

  • MDL number MFCD19381840
  • Cas Number : 186303-55-9
  • Molecular Weight : 426.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8344

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8344
Availability details for
Quantity
  • Availability
  • Add to Cart