COMH93D5EB62 Combi-Blocks, Inc.

[(5-Bromopyridin-2-yl)methyl](2-phenylethyl)amine

  • MDL number MFCD19424468
  • Cas Number : 1408397-61-4
  • Molecular Weight : 291.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8362
Availability details for
Quantity
  • Availability
  • Add to Cart