COMH93D5EB78 Combi-Blocks, Inc.

1-(3-Iodopropyl)-3-phenoxybenzene

  • MDL number MFCD19441075
  • Cas Number : 157126-72-2
  • Molecular Weight : 338.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8386

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8386
Availability details for
Quantity
  • Availability
  • Add to Cart