COMH93D5EC2A Combi-Blocks, Inc.

2-Hydroxybutyl methacrylate

  • MDL number MFCD19707078
  • Cas Number : 13159-51-8
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8610

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8610
Availability details for
Quantity
  • Availability
  • Add to Cart