COMH93D5EC32 Combi-Blocks, Inc.

Ethyl imidazo[1,5-a]pyridine-7-carboxylate

  • MDL number MFCD19981647
  • Cas Number : 518979-75-4
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8618
Availability details for
Quantity
  • Availability
  • Add to Cart