COMH93D5EC3D Combi-Blocks, Inc.

Benzyl n-[(1r,3s)-3-fluorocyclopentyl]carbamate

  • MDL number MFCD20037400
  • Cas Number : 1932288-21-5
  • Molecular Weight : 237.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8631

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8631
Availability details for
Quantity
  • Availability
  • Add to Cart