COMH93D5EC41 Combi-Blocks, Inc.

2-(6-(4-Chlorophenyl)pyridin-2-yl)acetonitrile

  • MDL number MFCD20038568
  • Cas Number : 23168-76-5
  • Molecular Weight : 228.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8635

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8635
Availability details for
Quantity
  • Availability
  • Add to Cart