COMH93D5EC98 Combi-Blocks, Inc.

3-Bromo-6-methoxypyrazolo[1,5-a]pyrimidine

  • MDL number MFCD20258249
  • Cas Number : 1314893-99-6
  • Molecular Weight : 228.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8747

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8747
Availability details for
Quantity
  • Availability
  • Add to Cart